Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxamine dihydrochloride, Cell Culture Reagent, Alfa Aesar™

Click to view available options
Quantity:
2.5g
5g
10g
524-36-7
C8H14Cl2N2O2
241.112
MFCD00012808
HNWCOANXZNKMLR-UHFFFAOYSA-N
pyridoxamine dihydrochloride, 4-aminomethyl-5-hydroxymethyl-2-methylpyridin-3-ol dihydrochloride, pyridoxamine dichlorohydrate, pyridoxylamine dihydrochloride, 4-aminomethyl-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride, unii-yq8ncr7v0o, pyridoxamine 2hcl, yq8ncr7v0o, 2-methyl-3-hydroxy-4-aminomethyl-5-hydroxymethylpyridene dihydrochloride, pyridoxemine, dihydrochloride
10664
4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol;dihydrochloride
CC1=NC=C(C(=C1O)CN)CO.Cl.Cl
524-36-7
C8H14Cl2N2O2
241.112
MFCD00012808
HNWCOANXZNKMLR-UHFFFAOYSA-N
pyridoxamine dihydrochloride, 4-aminomethyl-5-hydroxymethyl-2-methylpyridin-3-ol dihydrochloride, pyridoxamine dichlorohydrate, pyridoxylamine dihydrochloride, 4-aminomethyl-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride, unii-yq8ncr7v0o, pyridoxamine 2hcl, yq8ncr7v0o, 2-methyl-3-hydroxy-4-aminomethyl-5-hydroxymethylpyridene dihydrochloride, pyridoxemine, dihydrochloride
10664
4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol;dihydrochloride
CC1=NC=C(C(=C1O)CN)CO.Cl.Cl
Melting Point 220°C to 224°C
Quantity 2.5g
Packaging Glass bottle
Beilstein 3632748
Solubility Information Soluble in DMSO,water,or methanol /n
Formula Weight 241.12
Physical Form Solid
Chemical Name or Material Pyridoxamine dihydrochloride

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-357-6

RTECSNumber : UV1230000

TSCA : No

Recommended Storage : Store at -20°C