Learn More
AdipoGenSupplier Diversity Partner PYRIDOXATIN(1MG) DFS Item
Manufacturer:  AdipoGenSupplier Diversity Partner AGCN20123M001
Pyridoxatin, AdipoGen Life Sciences. Chemical. CAS: 135529-30-5. Formula: C15H21NO3. MW: 263.3. Isolated from Albophoma yamanashiensis. Antibiotic. Free radical scavenger. Lipid peroxidation inhibitor. Antimicrobial activity against Candida albicans. Gelatinase A (MMP-2) inhibitor. Anticancer. Cytotoxic. DNA synthesis inhibitor.
Catalog No. 50-114-6304