Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxine Hydrochloride, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
5g
100g
Packaging:
Wide Mouth Amber Glass Bottle
58-56-0
C8H12ClNO3
205.638
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
6019
CHEBI:30961
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
CC1=NC=C(C(=C1O)CO)CO.Cl
58-56-0
C8H12ClNO3
205.638
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
6019
CHEBI:30961
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
CC1=NC=C(C(=C1O)CO)CO.Cl
Quantity 100g
Packaging Wide Mouth Amber Glass Bottle
Formula Weight 205.64
Grade FCC/USP
Chemical Name or Material Pyridoxine Hydrochloride