Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrimethamine, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
58-14-0
C12H13ClN4
248.71
WKSAUQYGYAYLPV-UHFFFAOYSA-N
5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine
CCC1=NC(N)=NC(N)=C1C1=CC=C(Cl)C=C1
58-14-0
C12H13ClN4
248.71
WKSAUQYGYAYLPV-UHFFFAOYSA-N
5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine
CCC1=NC(N)=NC(N)=C1C1=CC=C(Cl)C=C1

P1602, 58-14-0, C12H13ClN4

Spectrum™ Pyrimethamine, USP is a folic acid antagonist used for protozoal infections. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 238-242°C
Quantity 500 g
CAS Max % 1
Percent Purity 99 to 101%