Alfa Aesar™ Pyrimidine-5-boronic acid, 98%

Manufacturer: Alfa Aesar™ H52984MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52984MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109299-78-7
    Molecular Formula C4H5BN2O2
    Formula Weight 123.91
    MDL Number MFCD03002366
    Chemical Name or Material Pyrimidine-5-boronic acid

    Specifications

    Formula Weight 123.91
    Molecular Formula C4H5BN2O2
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point >300°C
    Quantity 250mg
    MDL Number MFCD03002366
    CAS 109299-78-7
    Health Hazard 1 Danger
    Chemical Name or Material Pyrimidine-5-boronic acid
    Assay 98%

    Description

    Applications
    Pyrimidine 5-boronic acid could be used in an analogous one-pot copper-catalyzed DBAD coupling and condensation. The widely utilized Suzuki-Miyaura coupling allows for the coupling of boronic acids with carbon halides and surrogates.

    Solubility
    Soluble in Ether, MeOH, DMSO, THF.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit