Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrogallic Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

87-66-1
C6H6O3
126.111
WQGWDDDVZFFDIG-UHFFFAOYSA-N
1057
CHEBI:16164
benzene-1,2,3-triol
C1=CC(=C(C(=C1)O)O)O

High quality chemicals for laboratory and specialized industrial use

Quantity 125g
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.45kg
Formula Weight 126.11
Chemical Name or Material Pyrogallic Acid
Grade Baker Analyzed™ ACS Reagent