Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrogallol, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
87-66-1
C6H6O3
126.11
WQGWDDDVZFFDIG-UHFFFAOYSA-N
pyrogallol,1,2,3-trihydroxybenzene,pyrogallic acid,1,2,3-benzenetriol,fouramine brown ap,fourrine pg,fourrine 85,pyro,c.i. oxidation base 32,piral
1057
CHEBI:16164
benzene-1,2,3-triol
C1=CC(=C(C(=C1)O)O)O
87-66-1
C6H6O3
126.11
WQGWDDDVZFFDIG-UHFFFAOYSA-N
pyrogallol,1,2,3-trihydroxybenzene,pyrogallic acid,1,2,3-benzenetriol,fouramine brown ap,fourrine pg,fourrine 85,pyro,c.i. oxidation base 32,piral
1057
CHEBI:16164
benzene-1,2,3-triol
C1=CC(=C(C(=C1)O)O)O
Sulfated Ash 0.1% max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Melting Point 131.0°C to 135.0°C
Color White
Quantity 500g
Boiling Point 309.0°C
Assay Percent Range 98.5% min. (HPLC)
Linear Formula C6H3(OH)3
Beilstein 06,1071
Merck Index 15,8112
Solubility Information Solubility in water: 400g/L (25°C). Other solubilities: 1 g/1.3 mL alcohol, 1 g/1.6 mL ether, slightly soluble in benzene, chloroform and, carbon disulfide
Formula Weight 126.11
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Pyrogallol
Show More Show Less

GHS H Statement
Harmful to aquatic life with long lasting effects.
Toxic if swallowed.
Suspected of causing genetic defects.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serio

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irr

GHS Signal Word: Danger

EINECSNumber : 201-762-9