Alfa Aesar™ Pyrogallol, ACS, 99%

Manufacturer: Alfa Aesar™ 4415209

Quantity 10g

 View more versions of this product

Catalog No. AA4415209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 87-66-1
    Molecular Formula C6H6O3
    Formula Weight 126.11
    MDL Number MFCD00002192
    UN Number UN2811
    Beilstein 907431
    Merck Index 14,8000
    Chemical Name or Material Pyrogallol
    Grade ACS Grade

    Specifications

    Formula Weight 126.11
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 87-66-1
    Beilstein 907431
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    TSCA TSCA
    Molecular Formula C6H6O3
    MDL Number MFCD00002192
    UN Number UN2811
    Density 1.45g/mL
    Merck Index 14,8000
    RTECS Number UX2800000
    Sensitivity Air & Light Sensitive
    Odor Odorless
    Melting Point 131° to 135°C
    Boiling Point 309°C
    Health Hazard 1 Warning
    Physical Form Powder
    Color White
    Health Hazard 2 GHS H Statement
    H341-H302-H312-H332
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Quantity 10g
    Chemical Name or Material Pyrogallol
    Assay 99%
    Grade ACS Grade

    GHS H Statement
    H341-H302-H312-H332
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P281-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit