Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrogallol, Crystal, Reagent, ACS, Spectrum™

Click to view available options
Quantity:
25 g
500 g
6 x 125 g
6 x 500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
87-66-1
C6H6O3
126.11
WQGWDDDVZFFDIG-UHFFFAOYSA-N
benzene-1,2,3-triol
OC1=CC=CC(O)=C1O
87-66-1
C6H6O3
126.11
WQGWDDDVZFFDIG-UHFFFAOYSA-N
benzene-1,2,3-triol
OC1=CC=CC(O)=C1O

P1460, 87-66-1, C6H3(OH)3

Spectrum™ Pyrogallol, Crystal, Reagent, ACS , also known as Pyrogallic acid, is one of three isomeric benzenetriols that is used for oxygen absorption in gas analysis as well as in hair dying. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Packaging Amber Glass Bottle
Quantity 500 g
Melting Point 131°C to 135°C
Ignition Residue 5e-005
CAS Max % 1
Grade ACS