Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrogallol, Crystal, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog Number 18606014
Click to view available options
Quantity:
25 g
500 g
6 x 125 g
6 x 500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
87-66-1
C6H6O3
126.11
MFCD00002192
WQGWDDDVZFFDIG-UHFFFAOYSA-N
benzene-1,2,3-triol
OC1=CC=CC(O)=C1O
87-66-1
C6H6O3
126.11
MFCD00002192
WQGWDDDVZFFDIG-UHFFFAOYSA-N
benzene-1,2,3-triol
OC1=CC=CC(O)=C1O

P1460, 87-66-1, C6H3(OH)3

Spectrum™ Chemical Pyrogallol, Crystal, Reagent, ACS , also known as Pyrogallic acid, is one of three isomeric benzenetriols that is used for oxygen absorption in gas analysis as well as in hair dying. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

CAS Max % 1
Melting Point 131°C to 135°C
Ignition Residue 5e-005
Packaging Amber Glass Bottle
Quantity 6 x 500 g
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit