Learn More
Pyromellitic acid, 96%, Thermo Scientific Chemicals
Description
Pyromellitic acid is an eluent used in anion chromatography and forms complexes with magnesium and calcium ions. It is used as a curing agent, as an intermediate for powder coating and a cross-linking agent for alkyd resin. It is involved in the synthesis of polyimide and pyromellitic octyl.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pyromellitic acid is an eluent used in anion chromatography and forms complexes with magnesium and calcium ions. It is used as a curing agent, as an intermediate for powder coating and a cross-linking agent for alkyd resin. It is involved in the synthesis of polyimide and pyromellitic octyl.
Solubility
Soluble in alcohol.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Melting Point | 281°C to 285°C |
Quantity | 100 g |
Merck Index | 14,8004 |
Solubility Information | Soluble in alcohol. |
Formula Weight | 254.15 |
Percent Purity | 96% |
Chemical Name or Material | Pyromellitic acid |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 201-879-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.