Learn More
Pyromellitic dianhydride, 97%, Thermo Scientific Chemicals
Description
Pyromellitic dianhydride is used as a monomer in the preparation of polyimide polymers such as kapton; thermoplastics such as polyesters, polyethers, plasticizers, and expoxy resins. It is involved in the preparation of high performance coatings due to its mechanical, thermal and oxidative stability properties. It acts as a curing agent for epoxy resins which find application in adhesives, coatings and molding powders. As an intermediate, it plays an important role in polyimide based materials like printed circuit boards, tape automated bonding and magnetic wire insulation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pyromellitic dianhydride is used as a monomer in the preparation of polyimide polymers such as kapton; thermoplastics such as polyesters, polyethers, plasticizers, and expoxy resins. It is involved in the preparation of high performance coatings due to its mechanical, thermal and oxidative stability properties. It acts as a curing agent for epoxy resins which find application in adhesives, coatings and molding powders. As an intermediate, it plays an important role in polyimide based materials like printed circuit boards, tape automated bonding and magnetic wire insulation.
Solubility
Soluble in dimethyl sulfoxide, acetone, chloroform, ethyl ether, n-hexane and benzene.
Notes
Moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols and strong oxidizing agents.
Specifications
Melting Point | 284°C to 288°C |
Density | 1.68 |
Boiling Point | 305°C to 310°C (30 mmHg) |
Flash Point | 380°C (716°F) |
Quantity | 100 g |
Beilstein | 213583 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in dimethyl sulfoxide,acetone,chloroform,ethyl ether,n-hexane and benzene. |
Formula Weight | 218.12 |
Percent Purity | 97% |
Show More |
Safety and Handling
Hazard Category | H317-H318-H334 |
Hazard Statement | GHS H Statement H334-H318-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes serious eye damage. May cause an allergic skin reaction. |
Precautionary Statement | P261-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P310-P333+P313-P342+P311-P363-P501c |
EINECSNumber | 201-898-9 |
RTECSNumber | DB9300000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.