Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyronin B, 50% dye content, ACROS Organics™

Click to view available options
Quantity:
5g
25g
1229438-47-4
C21H27Cl4FeN2O
521.11
1229438-47-4
C21H27Cl4FeN2O
521.11
Quantity 25g
Melting Point 174.0°C to 176.0°C
Beilstein 18,V,10,182
Merck Index 15,8118
Formula Weight 521.11
Chemical Name or Material Pyronin B