Learn More
Aat BioquestSupplier Diversity Partner Pyronin Y *CAS 92-32-0* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 61
Pyronin Y is widely used in combination with methyl green to selectively and differentially stain nucleic acids following polyacrylamide gel electrophoresis.
Catalog No. 50-195-2688