Alfa Aesar™ Pyrophosphoryl chloride

Manufacturer: Alfa Aesar™ 2236202

Quantity 0.5g

 View more versions of this product

Catalog No. AA2236202

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13498-14-1
    Formula Weight 251.76
    MDL Number MFCD00061542
    UN Number UN3264
    Synonym Phosphoryl chloride, pyro
    Chemical Name or Material Pyrophosphoryl chloride

    Specifications

    Formula Weight 251.76
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Odor Acrid
    MDL Number MFCD00061542
    CAS 13498-14-1
    Synonym Phosphoryl chloride, pyro
    Color Colorless to pale yellow
    Boiling Point 101°C (10mmHg)
    Health Hazard 1 Danger
    Physical Form Liquid
    UN Number UN3264
    Density 1.82g/mL
    Solubility Information Reacts violently with water
    Sensitivity Air & Moisture Sensitive
    Melting Point −17°C
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
    Quantity 0.5g
    Chemical Name or Material Pyrophosphoryl chloride

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit