Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PYROXD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558341
Description
Immunogen sequence: MCEVKKIYLQ DEFRILKKKS FTFPRDHKSV TADTEMWPVY VELTNEKIYG CDFIVSATGV TPNVEPFLHG NSFDLGEDGG LKVDDHMHTS LPDIYAAG Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.
The PYROXD1 gene encodes the protein pyridine nucleotide-disulphide oxidoreductase domain 1, which has a significant role in mitochondrial function and cellular redox homeostasis. This gene is involved in maintaining the balance of oxidative stress within cells and is crucial for the redox regulation of cellular components, impacting processes like energy metabolism and apoptosis. Mutations in PYROXD1 have been associated with early-onset myopathy, indicating its importance in muscle development and maintenance. The protein encoded by PYROXD1 is part of the oxidoreductase enzyme family that facilitates electron transfer, contributing to cellular defense against oxidative damage. Recent studies have linked PYROXD1 to various neuromuscular disorders, where disruptions in its function affect muscle fiber alignment and force production. Understanding its role in redox biology is crucial for developing therapeutic strategies to combat muscle pathologies and redox-related diseases.
Specifications
PYROXD1 | |
Polyclonal | |
Unconjugated | |
PYROXD1 | |
FLJ22028; pyridine nucleotide-disulfide oxidoreductase domain-containing protein 1; pyridine nucleotide-disulphide oxidoreductase domain 1; PYROXD1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79912 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WU10 | |
PYROXD1 | |
Recombinant protein corresponding to Human PYROXD1. Recombinant protein control fragment (Product #RP-102824). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction