Pyrrhotite, Gray grains, Alfa Aesar™

For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals

Manufacturer: Alfa Aesar 4265203

Quantity 1g

 View more versions of this product

Catalog No. AA4265203

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Physical Form Grains
    Molecular Formula Fe7S8
    MDL Number MFCD01868205
    Chemical Name or Material Pyrrhotite, Naturally occuring mineral

    Specifications

    Physical Form Grains
    Formula Weight 647.44
    Quantity 1g
    Color Gray
    Density 4.58-4.65g/cm3 at 20°C
    Molecular Formula Fe7S8
    MDL Number MFCD01868205
    Chemical Name or Material Pyrrhotite, Naturally occuring mineral

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit