Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrole-3-carboxylic acid hydrate, 95%, Acros Organics™

Click to view available options
Quantity:
500mg
2.5g
Packaging:
Glass bottle
336100-46-0
C5H5NO2·xH2O
111.1
CFVUWWPGGBWCTB-UHFFFAOYSA-N
pyrrole-3-carboxylic acid hydrate,1h-pyrrole-3-carboxylic acid hydrate,c5h5no2.h2o,1h-pyrrole-3-carboxylicacid, hydrate 1:?
45076181
1H-pyrrole-3-carboxylic acid;hydrate
C1=CNC=C1C(=O)O.O
336100-46-0
C5H5NO2·xH2O
111.1
CFVUWWPGGBWCTB-UHFFFAOYSA-N
pyrrole-3-carboxylic acid hydrate,1h-pyrrole-3-carboxylic acid hydrate,c5h5no2.h2o,1h-pyrrole-3-carboxylicacid, hydrate 1:?
45076181
1H-pyrrole-3-carboxylic acid;hydrate
C1=CNC=C1C(=O)O.O
Water 14% max.
Melting Point 144.0°C to 147.0°C
Color Beige to Brown
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 2.5g
Assay Percent Range 94% min. (HPLC)
Beilstein 22,II,17
Solubility Information Solubility in water: soluble. Other solubilities: soluble in benzene, diethyl ether, petrol. ether
Formula Weight 111.1
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material Pyrrole-3-carboxylic acid hydrate
Show More Show Less

GHS Signal Word: Warning