Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrolidine +99%, ACROS Organics™

Click to view available options
Quantity:
100mL
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
123-75-1
C4H9N
71.11
MFCD00005249
RWRDLPDLKQPQOW-UHFFFAOYSA-N
tetrahydropyrrole,tetrahydro pyrrole,azacyclopentane,azolidine,tetramethylenimine,butylenimine,perhydropyrrole,prolamine,1-azacyclopentane,tetramethyleneimine
31268
CHEBI:33135
pyrrolidine
C1CCNC1
123-75-1
C4H9N
71.11
MFCD00005249
RWRDLPDLKQPQOW-UHFFFAOYSA-N
tetrahydropyrrole,tetrahydro pyrrole,azacyclopentane,azolidine,tetramethylenimine,butylenimine,perhydropyrrole,prolamine,1-azacyclopentane,tetramethyleneimine
31268
CHEBI:33135
pyrrolidine
C1CCNC1
Viscosity 0.94 mPa.s (20°C)
Boiling Point 86.0°C to 88.0°C
Quantity 2.5L
Flash Point 3°C
Refractive Index 1.4421 to 1.4441
Melting Point -63.0°C
Packaging Glass bottle
Specific Gravity 0.866
Density 0.8660g/mL
Assay Percent Range 99+%
Beilstein 20,4
Fieser 01,972; 02,354; 03,240; 07,309; 08,428; 13,14
Merck Index 15,8129
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether and chloroform, soluble in most organic solvents
Formula Weight 71.11
Percent Purity 99+%
Chemical Name or Material Pyrrolidine, 99+%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 204-648-7

RTECSNumber : UX9650000

TSCA : TSCA