Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrrolidine Hydrochloride 98.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
25150-61-2
C4H9ClN
106.57
MFCD01687885
QTSCTZWOLHXHRB-UHFFFAOYSA-N
Pyrrolidinium Chloride
212848
pyrrolidine hydrochloridyl
[Cl].C1CCNC1
25150-61-2
C4H9ClN
106.57
MFCD01687885
QTSCTZWOLHXHRB-UHFFFAOYSA-N
Pyrrolidinium Chloride
212848
pyrrolidine hydrochloridyl
[Cl].C1CCNC1
Color White
Quantity 5g
Formula Weight 107.58
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material Pyrrolidine Hydrochloride