Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyruvate Dehydrogenase E2 Rabbit anti-Human, Mouse, Rat, Clone: 2Z7R10, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Pyruvate Dehydrogenase E2 |
---|---|
Clone | 2Z7R10 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Pyruvate Dehydrogenase E2 Monoclonal antibody specifically detects Pyruvate Dehydrogenase E2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Pyruvate Dehydrogenase E2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1737 | |
IgG | |
Affinity purified |
2Z7R10 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
70 kDa mitochondrial autoantigen of primary biliary cirrhosis, dihydrolipoamide S-acetyltransferase, DLTA, E2 component of pyruvate dehydrogenase complex, EC 2.3.1, M2 antigen complex 70 kDa subunit, PBC, PDC-E2EC 2.3.1.12, PDCE2mitochondrial | |
A synthetic peptide corresponding to a sequence within amino acids 548-647 of human Pyruvate Dehydrogenase E2 (P10515). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title