Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvic Acid Sodium Salt, ≥98.5%, Affymetrix/USB™

Click to view available options
Quantity:
10g
100g
Packaging:
Plastic bottle
113-24-6
C3H3NaO3
110.044
MFCD00002586
DAEPDZWVDSPTHF-UHFFFAOYSA-M
sodium pyruvate,pyruvic acid sodium salt,sodium 2-oxopropanoate,pyruvic acid, sodium salt,propanoic acid, 2-oxo-, sodium salt,sodium alpha-ketopropionate,pyruvate sodium,unii-pod38aif08,2-oxopropanoic acid sodium salt,pod38aif08
23662274
CHEBI:50144
sodium;2-oxopropanoate
CC(=O)C(=O)[O-].[Na+]
113-24-6
C3H3NaO3
110.044
MFCD00002586
DAEPDZWVDSPTHF-UHFFFAOYSA-M
sodium pyruvate,pyruvic acid sodium salt,sodium 2-oxopropanoate,pyruvic acid, sodium salt,propanoic acid, 2-oxo-, sodium salt,sodium alpha-ketopropionate,pyruvate sodium,unii-pod38aif08,2-oxopropanoic acid sodium salt,pod38aif08
23662274
CHEBI:50144
sodium;2-oxopropanoate
CC(=O)C(=O)[O-].[Na+]
Linear Formula CH3COCOONa
Formula Weight 110.04
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Packaging Plastic bottle
Boiling Point >300°C
Quantity 10g
Chemical Name or Material Pyruvic Acid Sodium Salt