Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvic Acid, Sodium Salt, +99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
113-24-6
C3H3NaO3
110.04
MFCD00002586
DAEPDZWVDSPTHF-UHFFFAOYSA-M
sodium pyruvate,pyruvic acid sodium salt,sodium 2-oxopropanoate,pyruvic acid, sodium salt,propanoic acid, 2-oxo-, sodium salt,sodium alpha-ketopropionate,pyruvate sodium,unii-pod38aif08,2-oxopropanoic acid sodium salt,pod38aif08
23662274
CHEBI:50144
sodium;2-oxopropanoate
CC(=O)C(=O)[O-].[Na+]
113-24-6
C3H3NaO3
110.04
MFCD00002586
DAEPDZWVDSPTHF-UHFFFAOYSA-M
sodium pyruvate,pyruvic acid sodium salt,sodium 2-oxopropanoate,pyruvic acid, sodium salt,propanoic acid, 2-oxo-, sodium salt,sodium alpha-ketopropionate,pyruvate sodium,unii-pod38aif08,2-oxopropanoic acid sodium salt,pod38aif08
23662274
CHEBI:50144
sodium;2-oxopropanoate
CC(=O)C(=O)[O-].[Na+]
Appearance (10% soln. in water) clear colorless to slightly yellow
pH 5.5 to 6.5 (10% soln. at 20°C)
Loss on Drying 0.5% max.
Packaging Plastic bottle
Color Colorless to Yellow
Quantity 500g
Infrared Spectrum Authentic
Melting Point >300.0°C
Assay Percent Range 99+%
Linear Formula CH3COCO2Na
Beilstein 03, 608
Solubility Information Solubility in water: 47 g/100 mL (20°C).
Formula Weight 110.04
Physical Form Solution
Percent Purity ≥99%
Chemical Name or Material Pyruvic acid, sodium salt
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning