Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Pyruvic aldehyde, 35-45% w/w aq. soln

Click to view available options
Quantity:
25g
100g
500g
78-98-8
C3H4O2
72.063
MFCD00006960
AIJULSRZWUXGPQ-UHFFFAOYSA-N
methylglyoxal,pyruvaldehyde,pyruvic aldehyde,acetylformaldehyde,acetylformyl,propanal, 2-oxo,2-ketopropionaldehyde,propanedione,propanolone,methyl glyoxal
880
CHEBI:17158
2-oxopropanal
CC(=O)C=O
78-98-8
C3H4O2
72.063
MFCD00006960
AIJULSRZWUXGPQ-UHFFFAOYSA-N
methylglyoxal,pyruvaldehyde,pyruvic aldehyde,acetylformaldehyde,acetylformyl,propanal, 2-oxo,2-ketopropionaldehyde,propanedione,propanolone,methyl glyoxal
880
CHEBI:17158
2-oxopropanal
CC(=O)C=O
Applications
Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.

Solubility
Soluble in DMSO, water.

Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
Concentration or Composition (by Analyte or Components) 35 to 45% w/w aq. soln
Sensitivity Air Sensitive
Refractive Index 1.405
Density 1.2
Quantity 25g
Linear Formula CH3COCHO
Beilstein 906750
Merck Index 14,6081
Solubility Information Soluble in DMSO,water.
Formula Weight 72.06
Chemical Name or Material Pyruvic aldehyde
Show More Show Less

GHS H Statement
H301-H341-H315-H319-H335
Toxic if swallowed.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P370+P378q-P501c

H227-H302-H317-H318-H341

EINECSNumber : 201-164-8

RTECSNumber : UZ0700000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon