Learn More
Pyruvic aldehyde, 35-45% w/w aq. soln, Thermo Scientific Chemicals
Description
Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.
Solubility
Soluble in DMSO, water.
Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
Specifications
Density | 1.2 |
Refractive Index | 1.405 |
Linear Formula | CH3COCHO |
Quantity | 500 g |
Beilstein | 906750 |
Sensitivity | Air Sensitive |
Merck Index | 14,6081 |
Solubility Information | Soluble in DMSO,water. |
Formula Weight | 72.06 |
Concentration or Composition (by Analyte or Components) | 35 to 45% w/w aq. soln |
Show More |
Safety and Handling
Hazard Category | H227-H302-H317-H318-H341 |
Hazard Statement | GHS H Statement H301-H341-H315-H319-H335 Toxic if swallowed. Suspected of causing genetic defects. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P201-P202-P210-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P370+P378q-P501c |
EINECSNumber | 201-164-8 |
RTECSNumber | UZ0700000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.