Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvic aldehyde, 35-45 wt% solution in water, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
78-98-8, 7732-18-5
C3H4O2
72.06
MFCD00006960
AIJULSRZWUXGPQ-UHFFFAOYSA-N
methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal
880
CHEBI:17158
2-oxopropanal
CC(=O)C=O
78-98-8, 7732-18-5
C3H4O2
72.06
MFCD00006960
AIJULSRZWUXGPQ-UHFFFAOYSA-N
methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal
880
CHEBI:17158
2-oxopropanal
CC(=O)C=O
Packaging Glass bottle
Quantity 100g
Melting Point -20.0°C
Flash Point >71°C
Specific Gravity 1.2
Refractive Index 1.4209
Density 1.2000g/mL
pH 1 to 3
Boiling Point >72.0°C
Color Brown-Yellow to Yellow
Linear Formula CH3COCHO
Beilstein 01, 762
Merck Index 15, 6153
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, benzene and dmso
Formula Weight 72.06
Physical Form Solution
Percent Purity 35-45 wt.%
Chemical Name or Material Pyruvic aldehyde, 35-45 wt.%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Causes serious eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Danger

EINECSNumber : 201-164-8

RTECSNumber : UZ0700000

TSCA : TSCA

Recommended Storage : 4°C