Alfa Aesar™ Pyruvic aldehyde dimethyl acetal, 97+%

Manufacturer: Alfa Aesar™ B2498822

Quantity 100g

 View more versions of this product

Catalog No. AAB2498822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6342-56-9
    Molecular Formula CH3COCH(OCH3)2
    Formula Weight 118.13
    MDL Number MFCD00008758
    UN Number UN1993
    Beilstein 1560557
    Synonym Methylglyoxal dimethyl acetal; 1,1-Dimethoxyacetone
    Chemical Name or Material Pyruvic aldehyde dimethyl acetal

    Specifications

    Formula Weight 118.13
    TSCA TSCA
    UN Number UN1993
    Melting Point −57°C
    Boiling Point 133° to 135°C
    Flash Point 34°C (93°F)
    Quantity 100g
    Molecular Formula CH3COCH(OCH3)2
    MDL Number MFCD00008758
    CAS 6342-56-9
    Beilstein 1560557
    Synonym Methylglyoxal dimethyl acetal; 1,1-Dimethoxyacetone
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material Pyruvic aldehyde dimethyl acetal
    Assay ≥97%

    Description

    Applications
    Pyruvic aldehyde dimethyl acetal was used in the synthesis of methylglyoxal via hydrolysis in the presence of H2SO4. It was used to investigate the effects of methylglyoxal-mediated glycation on the structure, thermal stability and enzyme activity of yeast enolase in vivo.

    Solubility
    Soluble in water. 350 g/L at 20°C.

    Notes
    Do not store together with acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit