Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Pyruvonitrile, tech. 90%

Click to view available options
Quantity:
1g
5g
631-57-2
C3H3NO
69.063
MFCD00001838
QLDHWVVRQCGZLE-UHFFFAOYSA-N
pyruvonitrile,propanenitrile, 2-oxo,oxopropionitrile,2-oxopropiononitrile,2-oxopropionitrile,oxypropionitrile,pyruvic acid nitrile,unii-ulo441649v,accn,.alpha.-oxopropionitrile
69430
acetyl cyanide
CC(=O)C#N
631-57-2
C3H3NO
69.063
MFCD00001838
QLDHWVVRQCGZLE-UHFFFAOYSA-N
pyruvonitrile,propanenitrile, 2-oxo,oxopropionitrile,2-oxopropiononitrile,2-oxopropionitrile,oxypropionitrile,pyruvic acid nitrile,unii-ulo441649v,accn,.alpha.-oxopropionitrile
69430
acetyl cyanide
CC(=O)C#N
Applications
Pyruvonitrile was used in the synthesis of cuprate-carbonyl π-complexes by reaction with Me2CuLi in THF.

Solubility
Soluble in ether and acetonitrile.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Quantity 5g
Boiling Point 92°C to 93°C
Density 0.974g/mL
Refractive Index 1.3755
Color Colorless to Brown
Linear Formula CH3COCN
UN Number UN3273
Beilstein 1737633
Formula Weight 69.06
Physical Form Liquid
Percent Purity 90%
Grade Technical
Chemical Name or Material Pyruvonitrile
Show More Show Less

GHS H Statement
H225-H300-H330-H315-H335
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal if inhaled.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: NITRILES, FLAMMABLE, TOXIC, N.O.S.

storageNote1 : 2° to 8°C