PYZD 4409, Tocris™

Ubiquitin-activating enzyme (E1) inhibitor; Blocks degradation of p53 and cyclin D3; Inhibits NF-κB activation and induces cell death associated with ER stress; Displays antitumor effects in a mouse model of leukemia

Manufacturer: R&D Systems 4994/50

Quantity 50mg

 View more versions of this product

Catalog No. 49-945-0

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 423148-78-1
    Molecular Formula C14H7ClFN3O5
    Formula Weight 351.67
    Synonym 1-(3-Chloro-4-fluorophenyl)-4-[(5-nitro-2-furanyl)methylene]-3, 5-pyrazolidinedione
    Chemical Name or Material PYZD 4409

    Specifications

    Formula Weight 351.67
    Quantity 50mg
    Molecular Formula C14H7ClFN3O5
    CAS 423148-78-1
    Synonym 1-(3-Chloro-4-fluorophenyl)-4-[(5-nitro-2-furanyl)methylene]-3, 5-pyrazolidinedione
    Recommended Storage Store at -20°C
    Chemical Name or Material PYZD 4409

    Store at -20°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit