Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QBC Diagnostics F.A.S.T. Prepackaged AFB Kits
Complete AFB staining systems
Supplier: QBC Diagnostics 427409
Includes: QBC F.A.S.T AFB Smear Kit
- F.A.S.T Auramine O Stain Kit (120mL)
- Quality control slides (5)
- F.A.S.T SureFocus Miscroscope Slides (432)
- F.A.S.T Sputum Cups (400)
- F.A.S.T Applicator Sticks (400)
- F.A.S.T Auramine O Stain Kit (120mL)
- Quality control slides (5)
- F.A.S.T SureFocus Miscroscope Slides (432)
- Bottles of F.A.S.T Sputum Digestion Solution (27)
Description
- Three minute staining
- Supplies for 400 tests
Specifications
F.A.S.T AFB Smear Stain Kit | |
120mL Auramine O stain kit, 5 quality control slides, 432 SureFocus miscroscope slides, 400 sputum cups, 400 applicator sticks | |
15° to 25°C |
400 Tests | |
Detect positive AFB (tuberculosis) sputum samples | |
Auramine O |
Safety and Handling
ShelfLife : 1 Year
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction