Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ Qdot™ 705 Streptavidin Conjugate Non-distribution product as customer accommodation.

Qdot™ 705 Streptavidin Conjugate

$197.00 - $642.00

Specifications

Color Brown
Regulatory Status For research use only
Storage Buffer 1M betaine, 50mM borate, pH 8.3 with 0.05% Sodium azide
Concentration 1μm
Format Suspension
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Q10163MP Non-distribution product as customer accommodation. View Documents Molecular Probes™
Q10163MP
50μL Each for $197.00
Add to cart
 
Q10161MP Non-distribution product as customer accommodation. View Documents Molecular Probes™
Q10161MP
200μL Each for $642.00
Add to cart
 
Description

Description

Qdot™ 705 Streptavidin Conjugate comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Qdot™ nanocrystal). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays, and are also used in purification workflows to achieve targeted fractionation. Qdot™ nanocrystal conjugates are provided as 1µM solutions.

Important Features of Qdot™ Streptavidin Conjugates:
  • Streptavidin Qdot™ 705 conjugate has emission maxima of ~705 nm
  • Approximately 5 to 10 streptavidins per Qdot™ nanocrystal
  • Extremely photostable and bright fluorescence
  • Efficiently excited with single-line excitation sources
  • Narrow emission, large Stokes shift
  • Available in multiple colors
  • Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more

    Properties of Qdot™ Nanocrystals
    The Qdot™ streptavidin conjugate is the size of a large macromolecule or protein (~15–20 nm) and represents the brightest class of streptavidin detection reagents. Qdot™ streptavidin conjugates are made from a nanometer-scale crystal of a semiconductor material (CdSe), which is coated with an additional semiconductor shell (ZnS) to improve the optical properties of the material. The Qdot™ 705 and Qdot™ 800 streptavidin conjugates, which include CdSeTe, are made in a similar fashion. This core-shell material is further coated with a polymer shell that allows the materials to be conjugated to biological molecules and to retain their optical properties.

    Other Fluorescent Conjugates of Streptavidin are Available
    We offer several other Qdot™ colors or try the Qdot™ Streptavidin Sampler Kit containing Qdot™ streptavidin conjugates in six colors. In addition to nanocrystal conjugates, we offer a wide range of streptavidins conjugated to Alexa Fluor™ dyes, Oregon Green™ dye, enzyme conjugates, and traditional fluorophores like Texas Red™ dye, fluorescein (FITC) and more.

    Find Biotinylated Conjugates
    We offer an extensive line of biotinylated conjugates for use in biotin-streptavidin detection strategies.
  • Use the primary antibody search tool to find biotinylated primary antibodies
  • Use the secondary antibody selector tool to find biotinylated secondary antibodies and biotinylated anti-dye and anti-hapten antibodies

    Blocking Endogenous Biotin
    Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our >Endogenous Biotin-Blocking Kit to minimize this interference.

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
  • Specifications

    Specifications

    Brown
    1M betaine, 50mM borate, pH 8.3 with 0.05% Sodium azide
    Suspension
    For research use only
    1μm

    For Research Use Only. Not for use in diagnostic procedures.

    Provide Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit