EVOS™ Qdot™ LED Light Cubes
Image fluorscence samples for specific wavelength
Manufacturer: EVOS™ AMEP4659
 View more versions of this product
Catalog No. 12-563-576
  Check Availability
Add to Cart
    Specifications
    Product Type Qdot 565 light cube
    For Use With (Equipment) EVOS™ FL Color Imaging System, FL Auto Imaging System, FL Imaging System
    Focus Distance Excitation/Emission: 445nm/565nm
    Type LED light cube
  • Description & Specifications
Related Products
Product Content Feedback