Learn More
EVOS™ Qdot™ LED Light Cubes Available on GSA/VA Contract for Federal Government customers only.
Image fluorscence samples for specific wavelength
Manufacturer:  EVOS™ AMEP4660
 View more versions of this product
Catalog No. 12-563-577
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
LED light cube
Qdot 585 light cube
EVOS™ FL Color Imaging System, FL Auto Imaging System, FL Imaging System
Excitation/Emission: 445nm/585 nm