EVOS™ Qdot™ LED Light Cubes
Image fluorscence samples for specific wavelength
Manufacturer: EVOS™ AMEP4665
 View more versions of this product
Catalog No. 12-563-581
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Product Type Qdot 800 light cube
    For Use With (Equipment) EVOS™ FL Color Imaging System, FL Auto Imaging System, FL Imaging System
    Focus Distance Excitation/Emission: 445nm/800nm
    Type LED light cube
  • Description & Specifications
Related Products
Product Content Feedback