Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QKI/Quaking Antibody (PCRP-QKI-2F10), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314072AF405
This item is not returnable.
View return policy
Description
QKI/Quaking Monoclonal antibody specifically detects QKI/Quaking in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
QKI/Quaking | |
Monoclonal | |
Alexa Fluor 405 | |
DKFZp586I0923, HKQ, homolog of mouse quaking QKI (KH domain RNA binding protein), Hqk, hqkI, protein quaking, QK, QK1, QK3, quaking homolog, KH domain RNA binding (mouse), RNA binding protein HQK | |
Recombinant full-length human QKI/Quaking protein (Uniprot: Q96PU8) | |
0.1 mL | |
Neuroscience | |
9444 | |
Store at 4°C in the dark. | |
IgG2c |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-QKI-2F10 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction