Learn More
QKI/Quaking Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: S147-6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242196V
Catalog No. NBP242196V
Add to cart
Description
Description
QKI/Quaking Monoclonal antibody specifically detects QKI/Quaking in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
QKI/Quaking
S147-6
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
QKI
Fusion protein amino acids 1-341 (full-length) of human QKI-5. Mouse: 100% identity (341/341 amino acids identical). Rat: 99% identity (339/341 amino acids identical) >90% identity with QKI-6, QKI-7 and QKI-7b.
Protein G purified
Neuroscience
Primary
9444
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
DKFZp586I0923, HKQ, homolog of mouse quaking QKI (KH domain RNA binding protein), Hqk, hqkI, protein quaking, QK, QK1, QK3, quaking homolog, KH domain RNA binding (mouse), RNA binding protein HQK
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Greater then 90% identity with QKI-6, QKI-7 and QKI-7b.Detects approx 36-38kDa.