Learn More
Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays
Utilize the precision, consistency and optimized performance of Thermo Scientific™ QMS™ TDM Assays, ready-to-use, liquid immunoassays.
Supplier: Thermo Scientific™ 0373589
Description
Accurately measure therapeutic drug levels in serum and plasma with the Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays. These liquid, ready-to-use assays are a convenient alternative to more expensive and time-consuming technologies. QMS (Quantitative Microsphere System) immunoassays have excellent precision and accuracy, optimized performance on a wide range of general chemistry analyzers and lot-to-lot consistency.
Features:- Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
- Validated parameters assure lab-to-lab reproducibility
- Superb reagent stability
- Excellent correlation to reference methods
- Optimal sensitivity, precision and dynamic range
- Low cross-reactivity with metabolites and commonly co-administered drugs
- Low interference from endogenous substances
Includes:
Two reagent system, 22, 22mL
Specifications
2°C to 8°C | |
Vancomycin | |
Reagent |
Vancomycin Assay | |
Plasma, Serum | |
22 x 22 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.