Learn More
Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators Available on GSA/VA Contract for Federal Government customers only.
Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ TDM Calibrators, offered in Levels A to F.
Manufacturer:  Thermo Scientific™ 0374678
 View more versions of this product
Catalog No. 23-279-107
  Check Availability
Add to cart
Specifications
Description
Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators, which employ advanced microparticle technology that is easy-to-use.
Reduces Labor
  • Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
  • Based on competitive inhibition immunoassay principle whereby free-drug in the sample competes for antibody binding sites with drug molecules coated onto uniform microparticles
Ready-to-use
  • Liquid stable and ready-to-use
  • Applicable to general chemistry analyzers
  • Validated parameters assure lab-to-lab reproducibility
  • Superb reagent stability
Optimal Sensitivity
  • Excellent correlation to reference methods
  • Low cross-reactivity with metabolites and commonly co-administered drugs
Description & Specifications
Specifications
Lidocaine Calibrators
Levels A-F, 1 x 1mL
Store at 2° to 8°C
Lidocaine
Plasma
Calibrator