Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ White Polypropylene SturdeeSeal PE Foam Lined Caps
Click to view available options
Neck Finish:
120-400
20-410
24-410
28-400
28-410
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
For Use With (Equipment):
Bottle
Wide mouth bottle
No. per Case:
144 Cs.
1500 Cs.
1650 Cs.
2000 Cs.
210 Cs.
2500 Cs.
2700 Cs.
288 Cs.
3000 Cs.
3750 Cs.
4300 Cs.
5000 Cs.
576 Cs.
608 Cs.
72 Cs.
7200 Cs.
760 Cs.
Packaging:
Bagged
Bulk
Description
- Can be used when good impact strength and pliability are required
- Resistant to many chemicals including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
- Available in ribbed style
- Available in bulk or bagged packing
Specifications
Closure Liner | Polyethylene Foam |
Closure Type | Ribbed |
For Use With (Equipment) | Wide mouth bottle |
Color | White |
Material | Polypropylene |
No. per Case | 1650 Cs. |
Packaging | Bulk |
Neck Finish | 58-400 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction