Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Qorpak™ White Polypropylene SturdeeSeal PE Foam Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Features excellent sealing properties
Manufacturer:  Qorpak™ CAP00359
 View more versions of this product
Catalog No. 02-991-271
Description
Description
  • Can be used when good impact strength and pliability are required
  • Resistant to many chemicals including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
  • Available in ribbed style
  • Available in bulk or bagged packing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
SturdeeSeal™ Polyethylene Foam
5000 Cs.
Wide mouth bottle
Ribbed
White
Bulk
33-400