Learn More
Qorpak 2mil Amber Open End Bags
Protects light-sensitive contents from ultraviolet damage. Qorpak 2mil Amber Open End Bags is used for safe transfer and storage of samples and parts.
Manufacturer: Qorpak BAG09861
Description
- Made of virgin polyethylene
- Available in various sizes
Specifications
9 x 3 in. (22.8 x 7.6cm) | |
Amber | |
22.8cm | |
3 in. |
2 mil | |
9 in. | |
LDPE (Low-density Polyethylene) | |
7.6cm |