Learn More
Qorpak™ 4mil Clear Zip Bags with Write-On Block
For safe transfer and storage of samples and parts. Qorpak™ 4mil Clear Zip Bags with Write-On Block provides large writing space for sample notation.
Manufacturer:  Qorpak™ BAG09970
 View more versions of this product
Catalog No. 02-992-975
Add to cart
Description
Description
  • Made of low-density polyethylene (LDPE)
  • Reclosable air tight sealing provides aditional protection to contents
Specifications
Specifications
8 x 5 in. (20.3 x 12.7cm)
20.3cm
5 in.
4 mil
Clear
8 in.
LDPE (Low-density Polyethylene)
12.7cm
Air-tight Seal