Learn More
Qorpak 4mil Clear Zip Bags with Write-On Block
For safe transfer and storage of samples and parts. Qorpak 4mil Clear Zip Bags with Write-On Block provides large writing space for sample notation.
Manufacturer: Qorpak BAG09958
Description
- Made of low-density polyethylene (LDPE)
- Reclosable air tight sealing provides aditional protection to contents
Specifications
5 in. | |
LDPE (Low-density Polyethylene) | |
7.6cm | |
Air-tight Seal | |
5 x 3 in. (12.7 x 7.6cm) |
12.7cm | |
3 in. | |
4 mil | |
Clear |