Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Amber Boston Round Bottles with Black Phenolic Hole Cap and PTFE/Silicone Septa

General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles with Black Phenolic Hole Cap and PTFE/Silicone Septa ensures product integrity for long-term storage.

Manufacturer:  Qorpak™ GLC06410

 View more versions of this product

Catalog No. 02-991-956


  Check Availability
Add to cart

Description

Description

  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum
Specifications

Specifications

4 oz., 120mL
4 oz.
Amber
Glass
160/Cs.
22-400
48mm
Screw
120mL
Phenolic
4.4 in.
Bulk Pack
Black
1.88 in.
112mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit