Learn More
Qorpak™ Amber Boston Round Bottles with Black Phenolic F217 and PTFE Cap Available on GSA/VA Contract for Federal Government customers only.
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles with Black Phenolic F217 and PTFE Cap ensures product integrity for long-term storage.
Manufacturer:  Qorpak™ GLC10584
 View more versions of this product
Catalog No. 02-992-230
  Check Availability
Add to cart
Specifications
Description
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum
Description & Specifications
Specifications
16 oz., 480mL
16 oz.
F217 and PTFE
6.6 in.
Bulk Pack
Black
2.95 in.
168mm
480mL
Phenolic
Amber
Glass
60/Cs.
28-400
75mm
Screw