Learn More
Qorpak™ Amber Boston Round Bottles with Black Phenolic F217 and PTFE Cap
General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles with Black Phenolic F217 and PTFE Cap ensures product integrity for long-term storage.
Manufacturer:  Qorpak™ GLC09675
 View more versions of this product
Catalog No. 02-992-694
  Check Availability
Add to cart
Description
Description
  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum
Specifications
Specifications
4 oz., 120mL
4 oz.
F217 and PTFE
4.4 in.
Bulk Pack
Black
1.88 in.
112mm
120mL
Phenolic
Amber
Glass
160/Cs.
22-400
48mm
Screw