Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Amber Boston Round Bottles, Cleaned for Semi-Volatiles Available on GSA/VA Contract for Federal Government customers only.

Protects from UV radiation

Manufacturer:  Qorpak™ GLC04934

 View more versions of this product

Catalog No. 02-992-439


  Check Availability
Add to cart

Description

Description

  • Precleaned for semi-volatiles such as pesticides, PCB's and metals
  • Cleaned and certified
  • Narrow screw neck opening with curved shoulders
  • Glass offers minimal chemical interference with contents
  • With black PP cap and PTFE disc attached

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Certifications

Meets U.S. EPA specifications and guidelines for contaminant-free sample containers

Specifications

Specifications

32 oz., 960mL
32 oz.
Polypropylene
3.7 in.
Glass
White
94mm
Screw
960mL
EPA
Amber
8.2 in.
20/Cs.
33-400
210mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit