Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Qorpak™ Kaptclean™ Amber Boston Round Bottles, Cleaned for Volatiles Available on GSA/VA Contract for Federal Government customers only.

General use bottles ideal for contents which are UV sensitive. Qorpak™ Kaptclean™ Amber Boston Round Bottles, Cleaned for Volatiles absorbs all wavelengths in the ultraviolet spectrum.

Manufacturer:  Qorpak™ GLC04902

 View more versions of this product

Catalog No. 02-993-093



Description

Description

  • Narrow screw neck opening with curved shoulders
  • Glass offers minimal chemical interference with contents
  • Bottles cleaned for volatiles which include benzene, chloroform, toluene and carbon tetrachloride
  • Supplied pre-assembled with caps attached

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

2 oz., 60mL
2 oz.
F217 and PTFE
1.53 in.
Glass
20-400
94mm
60mL
Thermoset
Amber
3.7 in.
Green
39mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit