Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Qorpak™ Amber Boston Round Bottles with Green PP Cap and PTFE/Silicone-Septa

General use bottles ideal when UV protection is important. Qorpak™ Amber Boston Round Bottles with Green PP Cap and PTFE/Silicone-Septa ensure integrity for long term storage and offer protection for UV-sensitive contents.

Manufacturer:  Qorpak™ GLC01955

 View more versions of this product

Catalog No. 02-992-553



Description

Description

  • Features narrow screw neck opening, curved shoulders, and round body
  • Glass offers minimal chemical interference with contents
  • Absorb all wavelengths in ultraviolet spectrum
Specifications

Specifications

8 oz., 240mL
8 oz.
Amber
5.43 in.
24/Cs.
24-400
138mm
240mL
Polypropylene
2.36 in.
Glass
Green
60mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit