Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Amber Boston Round Bottles with Polypropylene Cap and PTFE Disc

General use bottles ideal for UV sensitive contents. Qorpak™ Amber Boston Round Bottles with Polypropylene Cap and PTFE Disc ensures product integrity for long-term storage.

Manufacturer:  Qorpak™ GLC10236

 View more versions of this product

Catalog No. 02-993-259


  Check Availability
Add to cart

Description

Description

  • Features a narrow screw neck opening, curved shoulders, and a round body
  • Black or white PP cap with friction fit 0.015 in. PTFE disc attached
  • Glass and PTFE discs offers minimal chemical interference with contents
  • Amber color absorb all wavelengths in the ultraviolet spectrum
Specifications

Specifications

8 oz., 240mL
8 oz.
Amber
5.43 in.
96/Cs. (Bulk)
24-400
138mm
240mL
Polypropylene
2.36 in.
Glass
Black
60mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit